Merck
  • FK228 augmented temozolomide sensitivity in human glioma cells by blocking PI3K/AKT/mTOR signal pathways.
FK228 augmented temozolomide sensitivity in human glioma cells by blocking PI3K/AKT/mTOR signal pathways.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2016-09-30)
YiHan Wu, Li Dong, SaRuLa Bao, MeiLing Wang, YongLi Yun, RunXiu Zhu
ABSTRACT
Temozolomide is a novel cytotoxic agent currently used as first-line chemotherapy for glioblastoma multiforme (GBM). Romidepsin (FK228), a histone deacetylase inhibitor, is a promising new class of antineoplastic agent with the capacity to induce growth arrest and/or apoptosis of cancer cells. However, combination of the two drugs in glioma remains largely unknown. In the present study, we evaluated the combinatory effects of FK228 with TMZ in glioma, and its molecular mechanisms responsible for these effects. Glioma cell lines were treated with TMZ, FK228 or the combination of drugs. The resistance effect including cytotoxicity and apoptosis was determined in glioma cells, respectively. We further evaluated the effects of FK228 in the PI3K/Akt-signaling pathway in vitro. Mice engrafted with 5×106 LN382 cells were treated with TMZ, FK228 or the combination of two drugs, and tumor weights and volumes were measured, respectively. FK228 enhanced the cytotoxic effects of TMZ in glioma cells compared to vehicle-treated controls or each drug alone. The combination of FK228 and TMZ-induced apoptosis was demonstrated by increased expression of cleaved-Caspase 3, Bax, cleaved-PARP, and decreased Bcl-2 expression. Furthermore, the expression of key components of the PI3K/Akt-signaling pathway showed that combination of FK228 and TMZ block PI3K/Akt pathways in vitro. This block effect was also confirmed in vivo in mice models. Mice treated with both FK228 and TMZ drugs showed significantly reduced tumor weights and volumes, compared to each drug alone. Our results suggested that FK228 augmented temozolomide sensitivity in human glioma cells partially by blocking PI3K/AKT/mTOR signal pathways. It thus may provide a promising target for improving the therapeutic outcome of TMZ-resistant gliomas, although further studies will be needed.
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Romidepsin, ≥98% (HPLC)