Merck
  • Distinct subcellular location of the Ca2+-binding protein S100A1 differentially modulates Ca2+-cycling in ventricular rat cardiomyocytes.
Distinct subcellular location of the Ca2+-binding protein S100A1 differentially modulates Ca2+-cycling in ventricular rat cardiomyocytes.
Journal of cell science (2005-01-18)
Patrick Most, Melanie Boerries, Carmen Eicher, Christopher Schweda, Mirko Völkers, Thilo Wedel, Stefan Söllner, Hugo A Katus, Andrew Remppis, Ueli Aebi, Walter J Koch, Cora-Ann Schoenenberger
ABSTRACT
Calcium is a key regulator of cardiac function and is modulated through the Ca2+-sensor protein S100A1. S100 proteins are considered to exert both intracellular and extracellular functions on their target cells. Here we report the impact of an increased intracellular S100A1 protein level on Ca2+-homeostasis in neonatal ventricular cardiomyocytes in vitro. Specifically, we compare the effects of exogenously added recombinant S100A1 to those resulting from the overexpression of a transduced S100A1 gene. Extracellularly added S100A1 enhanced the Ca2+-transient amplitude in neonatal ventricular cardiomyocytes (NVCMs) through a marked decrease in intracellular diastolic Ca2+-concentrations ([Ca2+]i). The decrease in [Ca2+]i was independent of sarcoplasmic reticulum Ca2+-ATPase (SERCA2a) activity and was probably the result of an increased sarcolemmal Ca2+-extrusion through the sodium-calcium exchanger (NCX). At the same time the Ca2+-content of the sarcoplasmic reticulum (SR) decreased. These effects were dependent on the uptake of extracellularly added S100A1 protein and its subsequent routing to the endosomal compartment. Phospholipase C and protein kinase C, which are tightly associated with this subcellular compartment, were found to be activated by endocytosed S100A1. By contrast, adenoviral-mediated intracellular S100A1 overexpression enhanced the Ca2+-transient amplitude in NVCMs mainly through an increase in systolic [Ca2+]i. The increased Ca2+-load in the SR was based on an enhanced SERCA2a activity while NCX function was unaltered. Overexpressed S100A1 colocalized with SERCA2a and other Ca2+-regulatory proteins at the SR, whereas recombinant S100A1 protein that had been endocytosed did not colocalize with SR proteins. This study provides the first evidence that intracellular S100A1, depending on its subcellular location, modulates cardiac Ca2+-turnover via different Ca2+-regulatory proteins.