- MiR-324-5p Suppresses Hepatocellular Carcinoma Cell Invasion by Counteracting ECM Degradation through Post-Transcriptionally Downregulating ETS1 and SP1.
MiR-324-5p Suppresses Hepatocellular Carcinoma Cell Invasion by Counteracting ECM Degradation through Post-Transcriptionally Downregulating ETS1 and SP1.
Hepatocellular carcinoma (HCC) is one of the common malignancies, which is highly metastatic and the third common cause of cancer deaths in the world. The invasion and metastasis of cancer cells is a multistep and complex process which is mainly initiated by extracellular matrix (ECM) degradation. Aberrant expression of microRNA has been investigated in HCC and shown to play essential roles during HCC progression. In the present study, we found that microRNA-324-5p (miR-324-5p) was downregulated in both HCC cell lines and tissues. Ectopic miR-324-5p led to the reduction of HCC cells invasive and metastatic capacity, whereas inhibition of miR-324-5p promoted the invasion of HCC cells. Matrix metalloproteinase 2 (MMP2) and MMP9, the major regulators of ECM degradation, were found to be downregulated by ectopic miR-324-5p, while upregulated by miR-324-5p inhibitor. E26 transformation-specific 1 (ETS1) and Specificity protein 1 (SP1), both of which could modulate MMP2 and MMP9 expression and activity, were presented as the direct targets of and downregulated by miR-324-5p. Downregulation of ETS1 and SP1 mediated the inhibitory function of miR-324-5p on HCC migration and invasion. Our study demonstrates that miR-324-5p suppresses hepatocellular carcinoma cell invasion and might provide new clues to invasive HCC therapy.