Merck
  • Thalamus-derived molecules promote survival and dendritic growth of developing cortical neurons.
Thalamus-derived molecules promote survival and dendritic growth of developing cortical neurons.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2012-11-02)
Haruka Sato, Yuma Fukutani, Yuji Yamamoto, Eiichi Tatara, Makoto Takemoto, Kenji Shimamura, Nobuhiko Yamamoto
ABSTRACT
The mammalian neocortex is composed of various types of neurons that reflect its laminar and area structures. It has been suggested that not only intrinsic but also afferent-derived extrinsic factors are involved in neuronal differentiation during development. However, the role and molecular mechanism of such extrinsic factors are almost unknown. Here, we attempted to identify molecules that are expressed in the thalamus and affect cortical cell development. First, thalamus-specific molecules were sought by comparing gene expression profiles of the developing rat thalamus and cortex using microarrays, and by constructing a thalamus-enriched subtraction cDNA library. A systematic screening by in situ hybridization showed that several genes encoding extracellular molecules were strongly expressed in sensory thalamic nuclei. Exogenous and endogenous protein localization further demonstrated that two extracellular molecules, Neuritin-1 (NRN1) and VGF, were transported to thalamic axon terminals. Application of NRN1 and VGF to dissociated cell culture promoted the dendritic growth. An organotypic slice culture experiment further showed that the number of primary dendrites in multipolar stellate neurons increased in response to NRN1 and VGF, whereas dendritic growth of pyramidal neurons was not promoted. These molecules also increased neuronal survival of multipolar neurons. Taken together, these results suggest that the thalamus-specific molecules NRN1 and VGF play an important role in the dendritic growth and survival of cortical neurons in a cell type-specific manner.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Donkey Anti-Mouse IgG Antibody, Cy3 conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
Anti-Microtubule-Associated Protein 2 (MAP2) Antibody, Chemicon®, from rabbit