Merck
Concise three-component synthesis of defucogilvocarcin M.
Organic letters (2004-07-17)
Isao Takemura, Koreaki Imura, Takashi Matsumoto, Keisuke Suzuki
ABSTRACT
[reaction: see text] Concise synthesis of defucogilvocarcin M was achieved via the [2 + 2 + 2] approach to beta-phenylnaphthalene structure.