Merck
  • Discovery and structure-activity relationship studies of N6-benzoyladenine derivatives as novel BRD4 inhibitors.
Discovery and structure-activity relationship studies of N6-benzoyladenine derivatives as novel BRD4 inhibitors.
Bioorganic & medicinal chemistry (2015-02-14)
Tomomi Noguchi-Yachide, Taki Sakai, Yuichi Hashimoto, Takao Yamaguchi
ABSTRACT
Bromodomain and extra-terminal domain (BET) proteins are epigenetic readers that bind to acetylated lysines in histones. Among them, BRD4 is a candidate target molecule of therapeutic agents for diverse diseases, including cancer and inflammatory disease. As a part of our continuing structural development studies of thalidomide to obtain a broad spectrum of biological modifiers based on the 'multi-template' approach, in this work we focused on BRD4-inhibitory activity, and discovered that N6-benzoyladenine derivatives exhibit this activity. Structure-activity relationship studies led to N6-(2,4,5-trimethoxybenzoyl)adenine (29), which exhibits potent BRD4 bromodomain1 inhibitory activity with an IC50 value of 0.427μM. N6-Benzoyladenine appears to be a new chemical scaffold for development of BRD4 inhibitors.
Sigma-Aldrich
9-Methyladenine, 97%