Merck
  • The use of a [4 + 2] cycloaddition reaction for the preparation of a series of 'tethered' Ru(II)-diamine and aminoalcohol complexes.
The use of a [4 + 2] cycloaddition reaction for the preparation of a series of 'tethered' Ru(II)-diamine and aminoalcohol complexes.
Organic & biomolecular chemistry (2007-03-23)
Fung Kei Cheung, Aidan M Hayes, David J Morris, Martin Wills
ABSTRACT
A series of catalysts have been prepared for use in the asymmetric transfer hydrogenation of ketones. The complexes were prepared using a [4 + 2] cycloaddition reaction at a key step in the reaction sequence. This provides a means for the synthesis of catalysts with modifications at specific sites.
Sigma-Aldrich
Chloro(1,5-cyclooctadiene)rhodium(I) dimer, 98%