Merck
  • Smiles rearrangements in Ugi- and Passerini-type couplings: new multicomponent access to O- and N-arylamides.
Smiles rearrangements in Ugi- and Passerini-type couplings: new multicomponent access to O- and N-arylamides.
The Journal of organic chemistry (2007-04-26)
Laurent El Kaïm, Marie Gizolme, Laurence Grimaud, Julie Oble
ABSTRACT
The use of Smiles rearrangement in Ugi- and Passerini-type couplings with electron-deficient phenols allows very straightforward multicomponent formation of O-aryl- and N-arylamides. Best yields were observed with the highly activated o- and p-nitrophenols, salicylic derivatives giving adducts in lower yields. The scope of these new reactions is further increased by the successful couplings of heterocyclic phenols such as hydroxypyridines and hydroxypyrimidines.
Sigma-Aldrich
Benzyl isocyanide, 98%