Merck
  • Helicobacter pylori eradication increases telomere length in gastric mucosa.
Helicobacter pylori eradication increases telomere length in gastric mucosa.
Hepato-gastroenterology (2012-10-31)
Rahmi Aslan, Ahmet Bektas, Abdulkerim Bedir, Hasan Alacam, Melek Suzer Aslan, Rukiye Nar, Beytullah Yildirim, Ibrahim Goren, Ozgur Ecemis, Muge Ustaoglu, Fikret Goren, Ali Okuyucu
ABSTRACT
Our purpose in this study was to analyze telomere length and telomerase activity before and after eradication treatment in gastric mucosa in patients positive for H. pylori. There were two groups: a control group (n=17) and a study group (n=21). For H. pylori eradication, the patients were administrated proton pump inhibitor (PPI) + clarithromycin + amoxicillin or PPI + metronidazole + tetracycline + bismuth for 14 days. Telomere length was analyzed with RT-PCR and telomerase activity with PCR-ELISA on biopsy specimens from the antrum. The result p<0.05 was considered significant. Prior to eradication, there was no significant difference between telomere lengths of the patient and control groups (2481.2±1823 and 2958.9±1345.7 bp, p=0.11, respectively). The telomere length of the study group became longer after eradication (before 2481.2±1823bp, after 3766.3±1608.8bp, p=0.01). Telomerase activity was not detected in either the patient or the control group. An increase in telomere length was observed with H. pylori eradication. This finding may indicate the importance of H. pylori eradication to avoid the development of gastric cancer.
Product Description
Roche
High Pure PCR Template Preparation Kit, pkg of 100 purifications, suitable for DNA extraction
Roche
TeloTAGGG Telomere Length Assay, sufficient for ≤50 reactions, kit of 1 (15 components), suitable for cell culture