Merck
  • Linoleic acid stimulates [Ca2+]i increase in rat pancreatic beta-cells through both membrane receptor- and intracellular metabolite-mediated pathways.
Linoleic acid stimulates [Ca2+]i increase in rat pancreatic beta-cells through both membrane receptor- and intracellular metabolite-mediated pathways.
PloS one (2013-04-09)
Yufeng Zhao, Li Wang, Jianhua Qiu, Dingjun Zha, Qiang Sun, Chen Chen
ABSTRACT
The role of the free fatty acid (FFA) receptor and the intracellular metabolites of linoleic acid (LA) in LA-stimulated increase in cytosolic free calcium concentration ([Ca(2+)]i) was investigated. [Ca(2+)]i was measured using Fura-2 as indicator in rat pancreatic β-cells in primary culture. LA (20 µM for 2 min) stimulated a transient peak increase followed by a minor plateau increase in [Ca(2+)]i. Elongation of LA stimulation up to 10 min induced a strong and long-lasting elevation in [Ca(2+)]i. Activation of FFA receptors by the non-metabolic agonist GW9508 (40 µM for 10 min) resulted in an increase in [Ca(2+)]i similar to that of 2-min LA treatment. Inhibition of acyl-CoA synthetases by Triacsin C suppressed the strong and long-lasting increase in [Ca(2+)]i. The increase in [Ca(2+)]i induced by 2 min LA or GW9508 were fully eliminated by exhaustion of endoplasmic reticulum (ER) Ca(2+) stores or by inhibition of phospholipase C (PLC). Removal of extracellular Ca(2+) did not influence the transient peak increase in [Ca(2+)]i stimulated by 2 min LA or GW9508. The strong and long-lasting increase in [Ca(2+)]i induced by 10 min LA was only partially suppressed by extracellular Ca(2+) removal or thapsigargin pretreatment, whereas remaining elevation in [Ca(2+)]i was eliminated after exhaustion of mitochondrial Ca(2+) using triphenyltin. In conclusion, LA stimulates Ca(2+) release from ER through activation of the FFA receptor coupled to PLC and mobilizes mitochondrial Ca(2+) by intracellular metabolites in β-cells.
Sigma-Aldrich
GW9508, ≥98% (HPLC)