Merck
  • Intracellular kinetics of the androgen receptor shown by multimodal Image Correlation Spectroscopy (mICS).
Intracellular kinetics of the androgen receptor shown by multimodal Image Correlation Spectroscopy (mICS).
Scientific reports (2016-03-05)
Chi-Li Chiu, Katherin Patsch, Francesco Cutrale, Anjana Soundararajan, David B Agus, Scott E Fraser, Daniel Ruderman
ABSTRACT
The androgen receptor (AR) pathway plays a central role in prostate cancer (PCa) growth and progression and is a validated therapeutic target. In response to ligand binding AR translocates to the nucleus, though the molecular mechanism is not well understood. We therefore developed multimodal Image Correlation Spectroscopy (mICS) to measure anisotropic molecular motion across a live cell. We applied mICS to AR translocation dynamics to reveal its multimodal motion. By integrating fluorescence imaging methods we observed evidence for diffusion, confined movement, and binding of AR within both the cytoplasm and nucleus of PCa cells. Our findings suggest that in presence of cytoplasmic diffusion, the probability of AR crossing the nuclear membrane is an important factor in determining the AR distribution between cytoplasm and the nucleus, independent of functional microtubule transport. These findings may have implications for the future design of novel therapeutics targeting the AR pathway in PCa.