Merck
Labeling of DNA probes with a photoactivatable hapten.
Analytical biochemistry (1989-03-01)
G H Keller, D P Huang, M M Manak
ABSTRACT
A photoactivatable reagent for introducing haptens onto DNA probes has been prepared using a commercially available bifunctional linker arm reagent and amino-derivatized 2,4-dinitrophenyl (DNP). The resulting compound (photo-DNP) couples efficiently to DNA using an ordinary sunlamp. Under optimum conditions, about 7-23 DNP molecules per 1000 bases are incorporated into the DNA. Hybridization experiments demonstrate that as little as 1.5 x 10(5) copies of target DNA can be detected by filter hybridization with a photo-DNP-labeled probe and immunochemical detection.
Sigma-Aldrich
N-Succinimidyl N-(2,4-dinitrophenyl)-6-aminocaproate, ≥95% (HPLC)