Merck
  • Kinase activity and phosphorylation of the largest subunit of TFIIF transcription factor.
Kinase activity and phosphorylation of the largest subunit of TFIIF transcription factor.
The Journal of biological chemistry (1999-07-31)
M Rossignol, A Keriel, A Staub, J M Egly
ABSTRACT
The largest subunit of the human basal transcription factor TFIIFalpha (also called RAP74) was reported previously to be the target of some phospho/dephosphorylation process. We show that TFIIFalpha possesses a serine/threonine kinase activity, allowing an autophosphorylation of the two residues at position serine 385 and threonine 389. Mutation analysis strongly suggests that autophosphorylation of both sites regulates the transcription elongation process. Moreover we also evidence three additional phosphorylation sites located at positions 207-230, 271-283, and 335-344. These sites are phosphorylated by casein kinase II-like kinases and TAF(II)250, a component of TFIID.