Merck
Synthesis and utilization of saccharide intermediates.
Carbohydrate research (1993-10-04)
D Becker, N Galili
ABSTRACT
A new method has been developed for preparation of partially pivaloylated saccharides in one step from readily available starting materials. These intermediates were used in the synthesis of disaccharides and a glucosteroid.
Sigma-Aldrich
Trimethylacetyl chloride, 99%