Merck
  • Rational ligand design for the arylation of hindered primary amines guided by reaction progress kinetic analysis.
Rational ligand design for the arylation of hindered primary amines guided by reaction progress kinetic analysis.
Journal of the American Chemical Society (2015-02-05)
Paula Ruiz-Castillo, Donna G Blackmond, Stephen L Buchwald
ABSTRACT
We report the Pd-catalyzed arylation of very hindered α,α,α-trisubstituted primary amines. Kinetics-based mechanistic analysis and rational design have led to the development of two biarylphosphine ligands that allow the transformation to proceed with excellent efficiency. The process was effective in coupling a wide range of functionalized aryl and heteroaryl halides under mild conditions.
Sigma-Aldrich
(t-Bu)PhCPhos Pd G3, ≥95%
Pricing and availability is not currently available.
Sigma-Aldrich
(t-Bu)PhCPhos, 95%
Pricing and availability is not currently available.