Merck
  • Multiple mechanisms repress N-Bak mRNA translation in the healthy and apoptotic neurons.
Multiple mechanisms repress N-Bak mRNA translation in the healthy and apoptotic neurons.
Cell death & disease (2013-08-24)
M Jakobson, M Jakobson, O Llano, J Palgi, U Arumäe
ABSTRACT
N-Bak is a neuron-specific BH3-only splice variant of pro-apoptotic Bcl-2 family member Bak. We have shown that its mRNA is stable in the neurons, whereas the protein cannot be detected by antibodies, suggesting a strong translational arrest of the mRNA. Here we identify two regulatory elements in the N-Bak mRNA that significantly repress translation in the luciferase reporter assay: an upstream open reading frame in the 5'-untranslated region (UTR) and naturally spliced exon-exon junction downstream of the premature translation termination codon in the 3'UTR. We also show that N-Bak mRNA is stored in granular structures in the sympathetic neurons and stays in these granules during intrinsic apoptosis. Finally, we confirm the absence of N-Bak protein by quantitative mass spectrometry analysis in the healthy, apoptotic or stressed sympathetic and cortical neurons. We conclude that N-Bak mRNA is translationally repressed by multiple mechanisms, and the protein does not participate in the classical apoptosis or cellular stress response.
Sigma-Aldrich
Anti-Nerve Growth Factor Antibody, clone 27/21, azide free, clone 27/21, Chemicon®, from mouse
Roche
Anti-Digoxigenin-POD, Fab fragments, from sheep