Merck
  • Comparative Analysis of Two Gene-Targeting Approaches Challenges the Tumor-Suppressive Role of the Protein Kinase MK5/PRAK.
Comparative Analysis of Two Gene-Targeting Approaches Challenges the Tumor-Suppressive Role of the Protein Kinase MK5/PRAK.
PloS one (2015-08-22)
Natalia Ronkina, Claus Johansen, Lisa Bohlmann, Juri Lafera, Manoj B Menon, Christopher Tiedje, Kathrin Laaß, Benjamin E Turk, Lars Iversen, Alexey Kotlyarov, Matthias Gaestel
ABSTRACT
MK5 (MAPK-activated protein kinase 5) or PRAK (p38-regulated and -activated kinase) are alternative names for a serine/threonine protein kinase downstream to ERK3/4 and p38 MAPK. A previous gene targeting approach for MK5/PRAK (termed here MK5/PRAK-Δex8) revealed a seemingly tumor-suppressive role of MK5/PRAK in DMBA-induced one step skin carcinogenesis and Ras-induced transformation. Here we demonstrate that an alternative targeting strategy of MK5/PRAK (termed MK5/PRAK-Δex6) increased neither tumor incidence in the one step skin carcinogenesis model, nor Ras-induced transformation in primary cells. Interestingly, due to the targeting strategies and exon skipping both knockouts do not completely abolish the generation of MK5/PRAK protein, but express MK5/PRAK deletion mutants with different biochemical properties depending on the exon targeted: Targeting of exon 6 leads to expression of an unstable cytoplasmic catalytically inactive MK5/PRAK-Δex6 mutant while targeting of exon 8 results in a more stable nuclear MK5/PRAK-Δex8 mutant with residual catalytic activity. The different properties of the MK5/PRAK deletion mutants could be responsible for the observed discrepancy between the knockout strains and challenge the role of MK5/PRAK in p53-dependent tumor suppression. Further MK5/PRAK knockout and knock-in mouse strains will be necessary to assign a physiological function to MK5/PRAK in this model organism.
Sigma-Aldrich
MISSION® esiRNA, targeting human MAPKAPK5 (2)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Sodium trichloroacetate, 97%