Merck
  • Nature of foulants and fouling mechanism in the Protein A MabSelect resin cycled in a monoclonal antibody purification process.
Nature of foulants and fouling mechanism in the Protein A MabSelect resin cycled in a monoclonal antibody purification process.
Biotechnology and bioengineering (2015-07-18)
Shaojie Zhang, William Daniels, Jeffrey Salm, Judy Glynn, Joseph Martin, Christopher Gallo, Ranga Godavarti, Giorgio Carta
ABSTRACT
The composition and origin of foulants and their spatial distribution within the particles of the Protein A MabSelect resin cycled in a mAb purification process are determined using electron and confocal microscopy techniques with gold and fluorescently labeled protein probes that associate with the foulants. The results show that the foulants are primarily related to the mAb product, are heterogeneously dispersed both on the outer surface and in the interior of the resin beads, and accumulate only when loading the conditioned CHO cell culture supernatant. Insignificant accumulation is seen if the process is run with purified mAb or with the null cell culture supernatant. When bound to the Protein A ligand, the mAb responsible for the observed fouling behavior is shown to associate with BSA and α-lactalbumin. This property is exploited using labeled versions of these lipophilic proteins to assess the effectiveness of improved resin cleaning processes and to elucidate the fouling mechanism. Resin fouling for this mAb appears to be consistent with the occurrence of conformational changes that occur upon binding, which, in turn, facilitate association of lipophilic proteins with the mAb. Upon desorption at low pH, these destabilized mAb complexes are deposited on and within the resin growing with each cycle and eventually leading to significant degradation of process performance.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture