Merck
  • Isolation, structure, and surfactant properties of polysaccharides from Ulva lactuca L. from South China Sea.
Isolation, structure, and surfactant properties of polysaccharides from Ulva lactuca L. from South China Sea.
International journal of biological macromolecules (2015-06-01)
Hua Tian, Xueqiong Yin, Qinghuan Zeng, Li Zhu, Junhua Chen
ABSTRACT
Two polysaccharides (ULP1 and ULP2) were isolated through ultrasonic-assisted extraction from green seaweed Ulva lactuca L. which was collected from the South China Sea. The highest yield of 17.57% was obtained under the conditions of 2% NaOH, 90 °C, material/water mass ratio 1:80, liquid extraction 5h and subsequent ultrasound-assisted extraction 1h. The structure of ULPs were characterized with periodate oxidation followed by Smith degradation, (1)H NMR, (13)C NMR spectroscopy, FTIR, and GPC. The molecular weights of ULP1 and ULP2 were 189 kDa and 230 kDa, respectively. The structural characteristics of ULP1 and ULP2 were quite similar. They were composed of rhamnose, xylose, glucose, and glucuronic acid. The content of rhamnose, xylose, glucose, glucuronic acid, sulfate was 51.2%, 12.3%, 20.1%, 16.4%, 12.0% for ULP1, respectively, and 60.8%, 14.2%, 8.2%, 16.8%, 26.8%, respectively, for ULP2. Both ULP1 and ULP2 showed good surface activity. 5 mg/mL ULP1 (2.62×10(-2) mmol/L) decreased the water surface tension to 51.63 mN/m. The critical micellar concentration of ULP1 and ULP2 was 1.01 mg/mL (5.3×10(-3) mmol/L) and 1.14 mg/mL (5.0×10(-3) mmol/L), respectively.
Sigma-Aldrich
L-(+)-Arabinose, 99%
Sigma-Aldrich
L-(+)-Arabinose, 98%
Sigma-Aldrich
L-(+)-Arabinose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
L-(−)-Fucose, ≥98% (GC)
Sigma-Aldrich
L-(+)-Arabinose, ≥99% (GC)
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Xylose, BioXtra, ≥99% (GC)
Sigma-Aldrich
D-(+)-Xylose, ≥99% (GC)
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG