Merck
  • Modifying Choroidal Neovascularization Development with a Nutritional Supplement in Mice.
Modifying Choroidal Neovascularization Development with a Nutritional Supplement in Mice.
Nutrients (2015-07-15)
Alina Adriana Ivanescu, Patricia Fernández-Robredo, Henar Heras-Mulero, Luis Manuel Sádaba-Echarri, Laura García-García, Vanessa Fernández-García, Maite Moreno-Orduna, Aitor Redondo-Exposito, Sergio Recalde, Alfredo García-Layana
ABSTRACT
We examined the effect of nutritional supplements (modified Age Related Eye Disease Study (AREDS)-II formulation containing vitamins, minerals, lutein, resveratrol, and omega-3 fatty acids) on choroidal neovascularization (CNV). Supplements were administered alone and combined with intravitreal anti-VEGF in an early-CNV (diode laser-induced) murine model. Sixty mice were evenly divided into group V (oral vehicle, intravitreal saline), group S (oral supplement, intravitreal saline), group V + aVEGF (oral vehicle, intravitreal anti-VEGF), and group S + aVEGF (oral supplement, intravitreal anti-VEGF). Vehicle and nutritional supplements were administered daily for 38 days beginning 10 days before laser. Intravitreal injections were administered 48 h after laser. Fluorescein angiography (FA) and flat-mount CD31 staining evaluated leakage and CNV lesion area. Expression of VEGF, MMP-2 and MMP-9 activity, and NLRP3 were evaluated with RT-PCR, zymography, and western-blot. Leakage, CNV size, VEGF gene and protein expression were lower in groups V + aVEGF, S + aVEGF, and S than in V (all p < 0.05). Additionally, MMP-9 gene expression differed between groups S + aVEGF and V (p < 0.05) and MMP-9 activity was lower in S + aVEGF than in V and S (both p < 0.01). Levels of MMP-2 and NLRP3 were not significantly different between groups. Nutritional supplements either alone or combined with anti-VEGF may mitigate CNV development and inhibit retinal disease involving VEGF overexpression and CNV.
Sigma-Aldrich
Tropicamide, solid
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Xylazine, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri