Merck
  • Interleukin-17A promotes the growth of human germinal center derived non-Hodgkin B cell lymphoma.
Interleukin-17A promotes the growth of human germinal center derived non-Hodgkin B cell lymphoma.
Oncoimmunology (2015-10-10)
Elisa Ferretti, Emma Di Carlo, Emanuela Ognio, Carla Guarnotta, Francesco Bertoni, Anna Corcione, Ignazia Prigione, Giulio Fraternali-Orcioni, Domenico Ribatti, Jean Louis Ravetti, Maurilio Ponzoni, Claudio Tripodo, Vito Pistoia
ABSTRACT
Interleukin (IL)-17A belongs to IL-17 superfamily and binds the heterodimeric IL-17 receptor (R)(IL-17RA/IL-17RC). IL-17A promotes germinal center (GC) formation in mouse models of autoimmune or infectious diseases, but the role of IL-17A/IL-17AR complex in human neoplastic GC is unknown. In this study, we investigated expression and function of IL-17A/IL-17AR in the microenvironments of 44 B cell non-Hodgkin lymphomas (B-NHL) of GC origin (15 follicular lymphomas, 17 diffuse large B cells lymphomas and 12 Burkitt lymphomas) and 12 human tonsil GC. Furthermore, we investigated the role of IL-17A in two
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, NMR reference standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)