Merck
  • Electrochemical polymerization of pyrene derivatives on functionalized carbon nanotubes for pseudocapacitive electrodes.
Electrochemical polymerization of pyrene derivatives on functionalized carbon nanotubes for pseudocapacitive electrodes.
Nature communications (2015-05-07)
John C Bachman, Reza Kavian, Daniel J Graham, Dong Young Kim, Suguru Noda, Daniel G Nocera, Yang Shao-Horn, Seung Woo Lee
ABSTRACT
Electrochemical energy-storage devices have the potential to be clean and efficient, but their current cost and performance limit their use in numerous transportation and stationary applications. Many organic molecules are abundant, economical and electrochemically active; if selected correctly and rationally designed, these organic molecules offer a promising route to expand the applications of these energy-storage devices. In this study, polycyclic aromatic hydrocarbons are introduced within a functionalized few-walled carbon nanotube matrix to develop high-energy, high-power positive electrodes for pseudocapacitor applications. The reduction potential and capacity of various polycyclic aromatic hydrocarbons are correlated with their interaction with the functionalized few-walled carbon nanotube matrix, chemical configuration and electronic structure. These findings provide rational design criteria for nanostructured organic electrodes. When combined with lithium negative electrodes, these nanostructured organic electrodes exhibit energy densities of ∼350 Wh kg(-1)electrode at power densities of ∼10 kW kg(-1)electrode for over 10,000 cycles.
Sigma-Aldrich
Naphthalene, 99%
Sigma-Aldrich
1-Aminopyrene, 97%
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Pyrene, puriss. p.a., for fluorescence, ≥99.0% (GC)
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Perylene, sublimed grade, ≥99.5%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Perylene, ≥99%
Sigma-Aldrich
Phenanthrene, 98%
Sigma-Aldrich
1-Pyrenecarboxylic acid, 97%
Sigma-Aldrich
Phenanthrene, sublimed grade, ≥99.5%