Merck
  • Method development for impurity profiling in SFC: The selection of a dissimilar set of stationary phases.
Method development for impurity profiling in SFC: The selection of a dissimilar set of stationary phases.
Journal of pharmaceutical and biomedical analysis (2015-01-30)
Charlene Galea, Debby Mangelings, Yvan Vander Heyden
ABSTRACT
Supercritical fluid chromatography (SFC) is drawing considerable interest as separation technique in the pharmaceutical industry. The technique is already well established in chiral separations both analytically and on a preparative scale. The use of SFC as a technique for drug impurity profiling is examined here. To define starting conditions in method development for drug impurity profiling, a set of dissimilar stationary phases is screened in parallel. The possibility to select a set of dissimilar columns using the retention factors (k-values) for a set of 64 drugs measured on 27 columns in SFC was examined. Experiments were carried out at a back-pressure of 150 bar and 25 °C with a mobile phase consisting of CO2 and methanol with 0.1% isopropylamine (5-40% over 10 min) at a flow rate of 3 mL/min. These k-values were then used to calculate correlation coefficients on the one hand and to perform a principal component analysis on the other. The Kennard and Stone algorithm, besides dendrograms and correlation-coefficient colour maps were used to select a set of 6 dissimilar stationary phases. The stationary phase characterization results from this study were compared to those from previous studies found in the literature. Retention mechanisms for compounds possessing different properties were also evaluated. The dissimilarity of the selected subset of 6 stationary phases was validated using mixtures of compounds with similar properties and structures, as one can expect in a drug impurity profile.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Carvedilol, ≥98% (HPLC), solid
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Salsalate, ≥98% (HPLC)
Sigma-Aldrich
Pizotifen, ≥98% (HPLC)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Benzylamine, ReagentPlus®, 99%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
1,1-Dimethylbiguanide hydrochloride, 97%
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
4-Benzylphenol, 99%
Sigma-Aldrich
o-Terphenyl, 99%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Terazosin hydrochloride, ≥98% (TLC), powder
Sigma-Aldrich
Procaine hydrochloride, ≥97%
Sigma-Aldrich
Pindolol, ≥98% (TLC), powder
Sigma-Aldrich
Promethazine hydrochloride
Sigma-Aldrich
Nicardipine hydrochloride, powder, ≥98%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Diclofenac sodium salt
Sigma-Aldrich
Dopamine hydrochloride
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Desipramine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Isopropylamine, ≥97.0% (GC)
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture