Merck
  • Nuclear retention of full-length HTT RNA is mediated by splicing factors MBNL1 and U2AF65.
Nuclear retention of full-length HTT RNA is mediated by splicing factors MBNL1 and U2AF65.
Scientific reports (2015-07-29)
Xin Sun, Pan P Li, Shanshan Zhu, Rachael Cohen, Leonard O Marque, Christopher A Ross, Stefan M Pulst, Ho Yin Edwin Chan, Russell L Margolis, Dobrila D Rudnicki
ABSTRACT
Huntington's disease (HD) is caused by a CAG repeat expansion in the huntingtin (HTT) gene. Recent evidence suggests that HD is a consequence of multimodal, non-mutually exclusive mechanisms of pathogenesis that involve both HTT protein- and HTT RNA-triggered mechanisms. Here we provide further evidence for the role of expanded HTT (expHTT) RNA in HD by demonstrating that a fragment of expHTT is cytotoxic in the absence of any translation and that the extent of cytotoxicity is similar to the cytotoxicity of an expHTT protein fragment encoded by a transcript of similar length and with a similar repeat size. In addition, full-length (FL) expHTT is retained in the nucleus. Overexpression of the splicing factor muscleblind-like 1 (MBNL1) increases nuclear retention of expHTT and decreases the expression of expHTT protein in the cytosol. The splicing and nuclear export factor U2AF65 has the opposite effect, decreasing expHTT nuclear retention and increasing expression of expHTT protein. This suggests that MBNL1 and U2AF65 play a role in nuclear export of expHTT RNA.
Sigma-Aldrich
MISSION® esiRNA, targeting human MBNL1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mbnl1
Sigma-Aldrich
Anti-Polyglutamine-Expansion Diseases Marker Antibody, clone 5TF1-1C2, ascites fluid, clone 5TF1-1C2, Chemicon®
Sigma-Aldrich
Anti-Spinocerebellar Ataxia Type 3 Antibody, clone 1H9, ascites fluid, clone 1H9, Chemicon®
Sigma-Aldrich
Anti-Huntingtin Protein Antibody, a.a. 181-810, clone 1HU-4C8, ascites fluid, clone 1HU-4C8, Chemicon®