Merck
  • Preparation of (+)-trans-isoalliin and its isomers by chemical synthesis and RP-HPLC resolution.
Preparation of (+)-trans-isoalliin and its isomers by chemical synthesis and RP-HPLC resolution.
Journal of biomolecular techniques : JBT (2014-09-05)
Lasanthi Jayathilaka, Shalini Gupta, Jin-Sheng Huang, Jenny Lee, Bao-Shiang Lee
ABSTRACT
Naturally occurring (+)-trans-isoalliin, (R(C)R(S))-(+)-trans-S-1-propenyl-L-cysteine sulfoxide, is a major cysteine sulfoxide in onion. The importance of producing it synthetically to support further research is very well recognized. The (+)-trans-isoalliin is prepared by chemical synthesis and reversed-phase (RP)-HPLC. First, S-2-propenyl-L-cysteine (deoxyalliin) is formed from L-cysteine and allyl bromide, which is then isomerized to S-1-propenyl-L-cysteine (deoxyisoalliin) by a base-catalyzed reaction. A mixture of cis and trans forms of deoxyisoalliin is formed and separated by RP-HPLC. Oxidation of the trans form of deoxyisoalliin by H2O2 produces a mixture of (-)- and (+)-trans-isoalliin. Finally, RP-HPLC is used successfully in separating (-)- and (+)-trans-isoalliin, and hence, (+)-trans-isoalliin is synthesized for the first time in this study. In addition, the (±) diastereomers of cis-isoalliin are also separated and purified by RP-HPLC.
Sigma-Aldrich
trans-1-Bromo-1-propene, contains copper as stabilizer, 99%