Merck
  • Ribosomal proteins L5 and L11 co-operatively inactivate c-Myc via RNA-induced silencing complex.
Ribosomal proteins L5 and L11 co-operatively inactivate c-Myc via RNA-induced silencing complex.
Oncogene (2013-10-22)
J-M Liao, X Zhou, A Gatignol, H Lu
ABSTRACT
Oncogene MYC is highly expressed in many human cancers and functions as a global regulator of ribosome biogenesis. Previously, we reported that ribosomal protein (RP) L11 binds to c-Myc and inhibits its transcriptional activity in response to ribosomal stress. Here, we show that RPL5, co-operatively with RPL11, guides the RNA-induced silencing complex (RISC) to c-Myc mRNA and mediates the degradation of the mRNA, consequently leading to inhibition of c-Myc activity. Knocking down of RPL5 induced c-Myc expression at both mRNA and protein levels, whereas overexpression of RPL5 suppressed c-Myc expression and activity. Immunoprecipitation revealed that RPL5 binds to 3'UTR of c-Myc mRNA and two subunits of RISC, TRBP (HIV-1 TAR RNA-binding protein) and Ago2, mediating the targeting of c-Myc mRNA by miRNAs. Interestingly, RPL5 and RPL11 co-resided on c-Myc mRNA and suppressed c-Myc expression co-operatively. These findings uncover a mechanism by which these two RPs can co-operatively suppress c-Myc expression, allowing a tightly controlled ribosome biogenesis in cells.
Sigma-Aldrich
MISSION® esiRNA, targeting human NCOA6
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ncoa6
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ncoa7
Sigma-Aldrich
MISSION® esiRNA, targeting human TARBP2
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture