Merck
  • The development of diet-induced obesity and associated metabolic impairments in Dj-1 deficient mice.
The development of diet-induced obesity and associated metabolic impairments in Dj-1 deficient mice.
The Journal of nutritional biochemistry (2014-12-03)
Katrin Seyfarth, Gereon Poschmann, Jan Rozman, Tobias Fromme, Nadine Rink, Andreas Hofmann, Wolfgang Wurst, Kai Stühler, Martin Klingenspor
ABSTRACT
DJ-1 constitutes a ubiquitously expressed, oxidative stress-responsive protein with multiple functions. DJ-1 emerged as a candidate from our previous proteome analysis investigating alterations in the hypothalamus in three mouse strains differing in their susceptibility to diet-induced obesity (DIO). Validation studies demonstrated a high-fat diet (HFD)-induced shift in the DJ-1 isoform pattern in the hypothalamus and several other tissues of mice. Others found HFD-induced alterations in DJ-1 protein abundance in adipose tissue and pancreatic islets in wild-type rodents. Here, we investigated the gene-diet interaction by challenging Dj-1(-/-) mice with a HFD. We demonstrate that the development of diet-induced obesity (DIO) Dj-1(-/-) mice is according to wild-type mice with the exception of transient higher gains in fat mass at the expense of lean mass after 14 weeks of feeding.
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, meets USP testing specifications
Urea, European Pharmacopoeia (EP) Reference Standard
Supelco
Urea, analytical standard
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Urea, suitable for electrophoresis