Merck
  • ⁶⁸Ga-DOTA-TOC-PET/CT detects heart metastases from ileal neuroendocrine tumors.
⁶⁸Ga-DOTA-TOC-PET/CT detects heart metastases from ileal neuroendocrine tumors.
Endocrine (2013-11-26)
Jan Calissendorff, Anders Sundin, Henrik Falhammar
ABSTRACT
Metastases from ileal neuroendocrine tumors (NETs) to the myocardium are rare and generally seen in patients with widespread metastatic NET disease. The objectives of this investigation were to describe the frequency of intracardiac metastases in ileal NET patients examined by (68)Ga-DOTA-TOC-PET/CT and to describe the cases in detail. All (68)Ga-DOTA-TOC-PET/CT examinations performed at the Karolinska University Hospital since 2010 until April 2012 were reviewed. In all, 128 out of 337 examinations were in patients with ileal NETs. Four patients had seven myocardiac metastases, yielding a frequency of 4.3 % in patients with ileal NETs. One patient had cardiac surgery while three were treated with somatostatin analogs. The cardiac metastases did not affect the patients' activity of daily life. (68)Ga-DOTA-TOC-PET/CT is an established imaging modality in identifying cardiac metastases in ileal NETs. Prospective studies are needed to confirm the true clinical value of (68)Ga-DOTA-TOC-PET/CT in detecting cardiac metastases in both ileal and non-ileal NETs.
Sigma-Aldrich
Octreotide, ≥98% (HPLC)