Merck
  • Separation and determination of impurities in paracetamol, codeine and pitophenone in the presence of fenpiverinium in combined suppository dosage form.
Separation and determination of impurities in paracetamol, codeine and pitophenone in the presence of fenpiverinium in combined suppository dosage form.
Journal of pharmaceutical and biomedical analysis (2014-09-26)
Jiří Vojta, Pavel Hanzlík, Aleš Jedlička, Pavel Coufal
ABSTRACT
A new HPLC method for separation and determination of impurities in paracetamol, codeine phosphate hemihydrate and pitophenone hydrochloride in the presence of fenpiverinium bromide in combined suppository dosage form was developed and validated. The separation of paracetamol and its impurities 4-aminophenol, 4-nitrophenol, 4-chloracetanilid; codeine and its impurities methylcodeine, morphine, codeine dimer and 10-hydroxycodeine; pitophenone and its impurities 2-[4-[2-(1-piperidinyl)ethoxy]benzoyl] benzoic acid, 2-[4-[2-(1-piperidinyl)ethoxy]benzoyl]benzoic acid 2-(1-piperidinyl)-ethyl ester, methyl ester of 2-(4-hydroxybenzoyl) benzoic acid and fenpiverinium was achieved by using ion-pair reversed phase liquid chromatography with UV detection. Validation parameters such as the precision, accuracy, linearity, limit of detection (LOD), limit of quantification (LOQ) and robustness were verified for all the mentioned impurities of codeine phosphate hemihydrate and 4-aminophenol and 2-[4-[2-(1-piperidinyl)ethoxy]benzoyl] benzoic acid as the main degradation products of paracetamol and pitophenone hydrochloride, respectively. The described method was found to be useful for analysis of the stability samples and therefore suitable for routine purity testing of the drug product.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Supelco
Methanol, analytical standard
Sigma-Aldrich
3-(Benzyldimethylammonio)propanesulfonate, BioXtra, ≥99.0% (HPCE)
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, ≥98%
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, BioXtra
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-(4-hydroxybenzoyl)benzoic acid, AldrichCPR
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)