Merck
  • Study of the protective effects of nootropic agents against neuronal damage induced by amyloid-beta (fragment 25-35) in cultured hippocampal neurons.
Study of the protective effects of nootropic agents against neuronal damage induced by amyloid-beta (fragment 25-35) in cultured hippocampal neurons.
Pharmacological reports : PR (2015-02-26)
Krzysztof Sendrowski, Wojciech Sobaniec, Anna Stasiak-Barmuta, Piotr Sobaniec, Janusz Popko
ABSTRACT
Alzheimer's disease (AD) is a common neurodegenerative disorder, in which progressive neuron loss, mainly in the hippocampus, is observed. The critical events in the pathogenesis of AD are associated with accumulation of β-amyloid (Aβ) peptides in the brain. Deposits of Aβ initiate a neurotoxic "cascade" leading to apoptotic death of neurons. Aim of this study was to assess a putative neuroprotective effects of two nootropic drugs: piracetam (PIR) and levetiracetam (LEV) on Aβ-injured hippocampal neurons in culture. Primary cultures of rat's hippocampal neurons at 7 day in vitro were exposed to Aβ(25-35) in the presence or absence of nootropics in varied concentrations. Flow cytometry with Annexin V/PI staining was used for counting and establishing neurons as viable, necrotic or apoptotic. Additionally, release of lactate dehydrogenase (LDH) to the culture medium, as a marker of cell death, was evaluated. Aβ(25-35) caused concentration-dependent death of about one third number of hippocampal neurons, mainly through an apoptotic pathway. In drugs-containing cultures, number of neurons injured with 20 μM Aβ(25-35) was about one-third lesser for PIR and almost two-fold lesser for LEV. When 40 μM Aβ(25-35) was used, only LEV exerted beneficial neuroprotective action, while PIR was ineffective. Our results suggest the protective potential of both studied nootropics against Aβ-induced death of cultured hippocampal neurons with more powerful neuroprotective effects of LEV.
Sigma-Aldrich
Piracetam
Piracetam, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Levetiracetam, ≥98% (HPLC)
USP
Levetiracetam, United States Pharmacopeia (USP) Reference Standard
Levetiracetam, European Pharmacopoeia (EP) Reference Standard
Supelco
Levetiracetam, Pharmaceutical Secondary Standard; Certified Reference Material