Merck
  • Mitochondrial translocation and interaction of cofilin and Drp1 are required for erucin-induced mitochondrial fission and apoptosis.
Mitochondrial translocation and interaction of cofilin and Drp1 are required for erucin-induced mitochondrial fission and apoptosis.
Oncotarget (2015-01-18)
Guobing Li, Jing Zhou, Amit Budhraja, Xiaoye Hu, Yibiao Chen, Qi Cheng, Lei Liu, Ting Zhou, Ping Li, Ehu Liu, Ning Gao
ABSTRACT
Cofilin is a member of the actin-depolymerizing factor (ADF) family protein, which plays an essential role in regulation of the mitochondrial apoptosis. It remains unclear how cofilin regulates the mitochondrial apoptosis. Here, we report for the first time that natural compound 4-methylthiobutyl isothiocyanate (erucin) found in consumable cruciferous vegetables induces mitochondrial fission and apoptosis in human breast cancer cells through the mitochondrial translocation of cofilin. Importantly, cofilin regulates erucin-induced mitochondrial fission by interacting with dynamin-related protein (Drp1). Knockdown of cofilin or Drp1 markedly reduced erucin-mediated mitochondrial translocation and interaction of cofilin and Drp1, mitochondrial fission, and apoptosis. Only dephosphorylated cofilin (Ser 3) and Drp1 (Ser 637) are translocated to the mitochondria. Cofilin S3E and Drp1 S637D mutants, which mimick the phosphorylated forms, suppressed mitochondrial translocation, fission, and apoptosis. Moreover, both dephosphorylation and mitochondrial translocation of cofilin and Drp1 are dependent on ROCK1 activation. In vivo findings confirmed that erucin-mediated inhibition of tumor growth in a breast cancer cell xenograft mouse model is associated with the mitochondrial translocation of cofilin and Drp1, fission and apoptosis. Our study reveals a novel role of cofilin in regulation of mitochondrial fission and suggests erucin as a potential drug for treatment of breast cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting human DENR
Sigma-Aldrich
MISSION® esiRNA, targeting human ROCK1 (2)
Sigma-Aldrich
MISSION® esiRNA, targeting human DNM1L
Sigma-Aldrich
MISSION® esiRNA, targeting human UTRN
Sigma-Aldrich
MISSION® esiRNA, targeting human DAPK2
Sigma-Aldrich
MISSION® esiRNA, targeting human CRMP1