Merck
  • MicroRNA expression in inflamed and noninflamed gingival tissues from Japanese patients.
MicroRNA expression in inflamed and noninflamed gingival tissues from Japanese patients.
Journal of oral science (2014-12-17)
Yorimasa Ogata, Sari Matsui, Ayako Kato, Liming Zhou, Yohei Nakayama, Hideki Takai
ABSTRACT
Periodontitis is a chronic inflammatory disease caused by specific bacteria and viruses. Local, systemic, and environmental factors affect the rate of disease progression. Immune responses to bacterial products, and the subsequent production of inflammatory cytokines, are crucial in the destruction of periodontal tissue. MicroRNAs (miRNAs) are a class of small RNAs that control various cell processes by negatively regulating protein-coding genes. In this study, we compared miRNA expression in inflamed and noninflamed gingival tissues from Japanese dental patients. Total RNAs were isolated from inflamed and noninflamed gingival tissues. miRNA expression profiles were examined by an miRNA microarray, and the data were analyzed by GeneSpring GX, Ingenuity Pathways Analysis, and the TargetScan databases. Observed miRNA expression levels in inflamed gingiva were confirmed by real-time PCR. The three most overexpressed (by >2.72-fold) miRNAs were hsa-miR-150, hsa-miR-223, and hsa-miR-200b, and the three most underexpressed (by <0.39-fold) miRNAs were hsa-miR-379, hsa-miR-199a-5p, and hsa-miR-214. In IPA analysis, hsa-miR-150, hsa-miR-223, and hsa-miR-200b were associated with inflammatory disease, organismal injury, abnormalities, urological disease, and cancer. The present findings suggest that miRNAs are associated with chronic periodontitis lesions in Japanese.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis