Merck
  • Crystal structure of Schistosoma mansoni arginase, a potential drug target for the treatment of schistosomiasis.
Crystal structure of Schistosoma mansoni arginase, a potential drug target for the treatment of schistosomiasis.
Biochemistry (2014-07-10)
Yang Hai, Jennifer E Edwards, Michael C Van Zandt, Karl F Hoffmann, David W Christianson
ABSTRACT
The X-ray crystal structure of arginase from Schistosoma mansoni (SmARG) and the structures of its complexes with several amino acid inhibitors have been determined at atomic resolution. SmARG is a binuclear manganese metalloenzyme that catalyzes the hydrolysis of l-arginine to form l-ornithine and urea, and this enzyme is upregulated in all forms of the parasite that interact with the human host. Current hypotheses suggest that parasitic arginases could play a role in host immune evasion by depleting pools of substrate l-arginine that would otherwise be utilized for NO biosynthesis and NO-dependent processes in the immune response. Although the amino acid sequence of SmARG is only 42% identical with that of human arginase I, residues important for substrate binding and catalysis are strictly conserved. In general, classical amino acid inhibitors such as 2(S)-amino-6-boronohexanoic acid (ABH) tend to bind more weakly to SmARG than to human arginase I despite identical inhibitor binding modes in each enzyme active site. The identification of a patch on the enzyme surface capable of accommodating the additional Cα substitutent of an α,α-disubstituted amino acid inhibitor suggests that such inhibitors could exhibit higher affinity and biological activity. The structures of SmARG complexed with two different α,α-disubstituted derivatives of ABH are presented and provide a proof of concept for this approach in the enhancement of enzyme-inhibitor affinity.
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, BioUltra, for molecular biology, ≥99.0% (KT)
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, 99.99% trace metals basis
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, meets USP testing specifications
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, ACS reagent, ≥98%