Merck
  • High GPX1 expression promotes esophageal squamous cell carcinoma invasion, migration, proliferation and cisplatin-resistance but can be reduced by vitamin D.
High GPX1 expression promotes esophageal squamous cell carcinoma invasion, migration, proliferation and cisplatin-resistance but can be reduced by vitamin D.
International journal of clinical and experimental medicine (2014-10-31)
Xiangfeng Gan, Baishen Chen, Zhuojian Shen, Yeqing Liu, Haifeng Li, Xuan Xie, Xia Xu, Haigang Li, Zhiquan Huang, Ju Chen
ABSTRACT
Esophageal cancer is one of the most common cancers worldwide. Despite recent progress in the development of novel therapies, esophageal carcinoma remains an aggressive cancer associated with a poor prognosis. The glutathione peroxidase 1 (GPX1) gene located on chromosome 3p21.3 is associated with the cancer of several organs. According to available information, GPX1, a gene downstream of NF-κB, is considered to exert adverse effects on tumour progression and enhance malignancy in some cancers but has not been reported in esophageal cancer. It is also reported that vitamin D (Vit. D), a widely used drug in the clinical setting, could suppress GPX1 expression through the NF-κB pathway. Thus, it is speculated that Vit. D could reduce malignancy in esophageal cancer by altering the NF-κB pathway. In this study, we confirmed our speculation by finding that Vit. D, through the inhibition of GPX1, decreased the migratory, invasive and proliferative capabilities, as well as cisplatin resistance, in esophageal cancer cells. Furthermore, when invasion and migration were reduced in the GPX1-inhibited cells, the expression of urokinase type plasminogen activator (uPA) and matrix metalloproteinase-2 (MMP2) was also suppressed correspondingly. Therefore, we believe that, in esophageal cancer cells, the expression of GPX1 can promote invasion, migration, proliferation and cisplatin resistance, and Vit. D can reduce the associated malignancy through the NF-κB pathway. The Vit. D- and NF-κB-mediated decrease in GPX1 expression resulted in a decrease in MMP2- and uPA-mediated invasion and migration.
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
MISSION® esiRNA, targeting human GPX1
Cisplatin, European Pharmacopoeia (EP) Reference Standard