Merck
  • Habiterpenol, a novel abrogator of bleomycin-induced G2 arrest in Jurkat cells, produced by Phytohabitans suffuscus 3787_5.
Habiterpenol, a novel abrogator of bleomycin-induced G2 arrest in Jurkat cells, produced by Phytohabitans suffuscus 3787_5.
The Journal of antibiotics (2014-06-12)
Ryuji Uchida, Sayaka Yokota, Daisuke Matsuda, Atsuko Matsumoto, Susumu Iwamoto, Hideyuki Onodera, Yoko Takahashi, Hiroshi Tomoda
ABSTRACT
A small molecule named habiterpenol produced by actinomycete Phytohabitans suffuscus 3787_5 was found to abrogate bleomycin-induced G2 arrest in Jurkat cells. Habiterpenol showed no cytotoxic effect on Jurkat cells even at 273 μM; however, the compound inhibited bleomycin-induced G2 arrest in Jurkat cells with an IC50 value of 3.55 μM, while it showed no effect on colchicine-induced M arrest even at 273 μM. These results indicated that habiterpenol selectively abrogated bleomycin-induced G2 arrest in Jurkat cells.
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Potassium fluoride, anhydrous, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Potassium fluoride, ≥99.97% trace metals basis
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Potassium fluoride, BioUltra, ≥99.5% (F)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Sigma-Aldrich
Colchicine, BioReagent, suitable for plant cell culture, ≥95% (HPLC)
Colchicine, (European Pharmacopoeia (EP) Reference Standard)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)