Merck
  • Pharmacodynamic analysis of intravenous recombinant urate oxidase using an indirect pharmacological response model in healthy subjects.
Pharmacodynamic analysis of intravenous recombinant urate oxidase using an indirect pharmacological response model in healthy subjects.
Acta pharmacologica Sinica (2014-10-07)
Ning-fang Cai, Ze-neng Cheng, Ying Zi, Xi Luo, Xin Guo, Zhi Liu, Li-yun Zheng
ABSTRACT
Pharmacodynamic analysis of intravenous recombinant urate oxidase produced by Escherichia coli was performed in healthy subjects using a pharmacokinetic/pharmacodynamic (PK/PD) model. A randomized, single-blind, placebo-controlled study was performed in 40 healthy Chinese subjects (4 groups of 10 subjects each, placebo 4:1 ratio) who received infusions of uricase (single doses of 0.1, 0.2, and 0.3 mg/kg; multiple doses of 0.2 mg·kg(-1)·d(-1) for 7 d). PK profiles were determined through plasma uricase activity, and PD profiles were established using uric acid levels in plasma and urine. The plasma PD parameter was estimated as changes in plasma uric acid levels as the effect in the indirect response model. Adverse events were also monitored. A two-compartment PK model with constant iv input and first-order output was used to describe the kinetic process of plasma uricase. The low value (2.8 U/L) of drug concentration that achieved 50% of maximum effect (EC50) indicated that low plasma uricase concentrations were sufficient to produce pharmacological effects. A strong relationship (r(2)=0.9991) between the mean uric acid concentration in blood and the mean uric acid excretion rate in urine in the range of 11 to 30 h after single dosing was found. Infusions of uricase were well tolerated in all subjects. The PK/PD model predicted the effective dose to be 0.1 mg/kg in healthy subjects. The excretion rate of uric acid in urine may be used as a new index for pharmacological effects in further clinical trials.
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%