Merck
  • Prostaglandin E2 elicits greater bronchodilation than salbutamol in mouse intrapulmonary airways in lung slices.
Prostaglandin E2 elicits greater bronchodilation than salbutamol in mouse intrapulmonary airways in lung slices.
Pulmonary pharmacology & therapeutics (2013-12-03)
M FitzPatrick, C Donovan, J E Bourke
ABSTRACT
Current asthma therapy may not adequately target contraction of smaller intrapulmonary airways, which are a major site of airway obstruction and inflammation. The aim of this study was to characterise responses of mouse intrapulmonary airways to prostaglandin E(2) (PGE(2)) and compare its dilator efficacy with the β(2)-adrenoceptor agonist salbutamol in situ, using lung slices. Lung slices (150 μm) were prepared from male Balb/C mice. Changes in intrapulmonary airway lumen area were recorded and analysed by phase-contrast microscopy. Relaxation to PGE(2) and salbutamol were assessed following various levels of pre-contraction with methacholine, serotonin or endothelin-1, as well as following overnight incubation with PGE(2) or salbutamol. The mechanism of PGE(2)-mediated relaxation was explored using selective EP antagonists (EP(1/2) AH6809; EP(4) L-161982) and Ca(2+)-permeabilized slices, where airway responses are due to regulation of Ca(2+)-sensitivity alone. PGE2 elicited EP(1/2)-mediated relaxation of intrapulmonary airways. PGE(2) was more potent than salbutamol in opposing submaximal pre-contraction to all constrictors tested, and only PGE(2) opposed maximal pre-contraction with endothelin-1. Relaxation to PGE(2) was maintained when contraction to methacholine was mediated via increased Ca(2+)-sensitivity alone. PGE(2) was less sensitive to homologous or heterologous desensitization of its receptors than salbutamol. The greater efficacy and potency of PGE(2) compared to salbutamol in mouse intrapulmonary airways supports further investigation of the mechanisms underlying this improved dilator responsiveness for the treatment of severe asthma.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
SAFC
HEPES
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
SAFC
HEPES
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Streptomycin sulfate salt, powder
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Caffeine, European Pharmacopoeia (EP) Reference Standard
Dinoprostone, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)