Merck
  • Fabrication, characterization, and biocompatibility of ethyl cellulose/carbonated hydroxyapatite composite coatings on Ti6Al4V.
Fabrication, characterization, and biocompatibility of ethyl cellulose/carbonated hydroxyapatite composite coatings on Ti6Al4V.
Journal of materials science. Materials in medicine (2014-05-27)
Bo Tian, Sha Tang, Yang Li, Teng Long, Xin-Hua Qu, De-Gang Yu, Ya-Jun Guo, Ya-Ping Guo, Zhen-An Zhu
ABSTRACT
In order to improve the biocompatibility of metallic implants, bioactive components are often used as coatings so that a real bond with the surrounding bone tissue can be formed. We prepared ethyl cellulose/carbonated hydroxyapatite composite coatings (ECHCs) on Ti6Al4V substrates with carbonated hydroxyapatite coatings (CHACs) without ethyl cellulose as controls. The inorganic constituent on the CHACs and ECHCs is calcium-deficient carbonated hydroxyapatite with a flaky texture and a low degree of crystallinity. The flaky carbonated hydroxyapatite plates aggregate to form macropores with an aperture size of around 0.5-2.0 μm. The presence of ethyl cellulose provides superior morphology, contact angle, and biocompatibility characteristics. In comparison to CHACs, ECHCs exhibit a smoother, crack-free surface because the cracks are filled by ethyl cellulose. Moreover, the contact angle of ECHCs is 37.3°, greater than that of CHACs (13.0°). Surface biocompatibility was investigated by using human bone mesenchymal stem cells (hBMSCs). The attachment, spreadability, viability and proliferation of hBMSCs on ECHCs are superior to those on CHACs. Thus, the crack-free ECHCs have excellent biocompatibility and are appropriate for use as biological implants.
Product Description
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Hydrofluoric acid, 48 wt. % in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrofluoric acid, ACS reagent, 48%
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard