Merck
  • A tris (2-carboxyethyl) phosphine (TCEP) related cleavage on cysteine-containing proteins.
A tris (2-carboxyethyl) phosphine (TCEP) related cleavage on cysteine-containing proteins.
Journal of the American Society for Mass Spectrometry (2010-03-02)
Peiran Liu, Brian W O'Mara, Bethanne M Warrack, Wei Wu, Yunping Huang, Yihong Zhang, Rulin Zhao, Mei Lin, Michael S Ackerman, Peter K Hocknell, Guodong Chen, Li Tao, Siegfried Rieble, Jack Wang, David B Wang-Iverson, Adrienne A Tymiak, Michael J Grace, Reb J Russell
ABSTRACT
Introduced in the late 1980s as a reducing reagent, Tris (2-carboxyethyl) phosphine (TCEP) has now become one of the most widely used protein reductants. To date, only a few studies on its side reactions have been published. We report the observation of a side reaction that cleaves protein backbones under mild conditions by fracturing the cysteine residues, thus generating heterogeneous peptides containing different moieties from the fractured cysteine. The peptide products were analyzed by high performance liquid chromatography and tandem mass spectrometry (LC/MS/MS). Peptides with a primary amine and a carboxylic acid as termini were observed, and others were found to contain amidated or formamidated carboxy termini, or formylated or glyoxylic amino termini. Formamidation of the carboxy terminus and the formation of glyoxylic amino terminus were unexpected reactions since both involve breaking of carbon-carbon bonds in cysteine.
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium citrate tribasic dihydrate, for molecular biology, ≥99%
Supelco
Tris(2-carboxyethyl)phosphine hydrochloride solution, 0.5 M, pH 7.0(aqueous solution; pH was adjusted with ammonium hydroxide)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%