Merck
  • Lack of association of the TP53BP1 Glu353Asp polymorphism with risk of cancer: a systematic review and meta-analysis.
Lack of association of the TP53BP1 Glu353Asp polymorphism with risk of cancer: a systematic review and meta-analysis.
PloS one (2014-03-08)
Lei Liu, Jinghua Jiao, Yu Wang, Dong Zhang, Jingyang Wu, Desheng Huang
ABSTRACT
The TP53BP1 gene may be involved in the development of cancer through disrupting DNA repair. However, studies investigating the relationship between TP53BP1 Glu353Asp (rs560191) polymorphism and cancer yielded contradictory and inconclusive outcomes. In order to realize these ambiguous findings, a meta-analysis was performed to assess the association between the TP53BP1 Glu353Asp (rs560191) polymorphism and susceptibility to cancer. We conducted a search of all English reports on studies for the association between the TP53BP1 Asp353Glu (rs560191) polymorphism and susceptibility to cancer using Medline, the Cochrane Library, EMbase, Web of Science, Google (scholar), and all Chinese reports were identified manually and on-line using CBMDisc, Chongqing VIP database, and CNKI database. The strict selection criteria and exclusion criteria were determined, and odds ratios (ORs) with 95% confidence intervals (CIs) were used to assess the strength of associations. The fixed or random effect model was selected based on the heterogeneity test among studies. Publication bias was estimated using funnel plots and Egger's regression test. A total of seven studies were included in the meta-analysis including 3,213 cases and 3,849 controls. The results indicated that the Glu353Asp (rs560191) polymorphism in TP53BP1 gene had no association with cancer risk for all genetic models. In the subgroup analysis, the results suggested that Glu353Asp polymorphism was not associated with the risk of cancer according to ethnicity, cancer type, genotyping method, adjusted with control or not, HWE and quality score. This meta-analysis suggested that the Glu353Asp (rs560191) polymorphism in TP53BP1 gene was not associated with risk of cancer.