Merck
  • Effect of sedative premedication on patient experience after general anesthesia: a randomized clinical trial.
Effect of sedative premedication on patient experience after general anesthesia: a randomized clinical trial.
JAMA (2015-03-04)
Axel Maurice-Szamburski, Pascal Auquier, Véronique Viarre-Oreal, Philippe Cuvillon, Michel Carles, Jacques Ripart, Stéphane Honore, Thibaut Triglia, Anderson Loundou, Marc Leone, Nicolas Bruder
ABSTRACT
Sedative premedication is widely administered before surgery, but little clinical evidence supports its use. To assess the efficacy of sedative premedication on perioperative patient experience. A randomized clinical trial, the PremedX study, enrolled 1062 adult patients who were younger than 70 years and had been scheduled for various elective surgeries under general anesthesia at 5 French teaching hospitals (in Marseille, Montpellier, Nimes, and Nice) between January 2013 and June 2014. Neurosurgery, obstetrical, cardiac, and outpatient surgery were excluded. Patients were randomized to 3 groups of 354 participants each to receive 2.5 mg of lorazepam, no premedication, or placebo. The primary outcome was perioperative patient experience assessed 24 hours after surgery with a validated questionnaire (Evaluation du Vécu de l'Anesthésie Generale; EVAN-G) describing 6 domains of satisfaction and a global index (score range, 0-100; high scores represent high satisfaction); secondary outcomes included time to extubation and early cognitive recovery. A subgroup analysis was planned a priori in patients with a high level of preoperative anxiety. Premedication with lorazepam did not improve the EVAN-G mean global index for overall level of patient satisfaction (72 [95% CI, 70-73]; n = 330) compared with no premedication (73 [95% CI, 71-74]; n = 319) or placebo (71 [95% CI, 70-73]; n = 322) (P = .38). Among patients with heightened preoperative anxiety, there were no significant differences found in the EVAN-G mean global index between the lorazepam group (68 [95% CI, 65-72]; n = 87) and the no premedication group (73 [95% CI, 69-77]; n = 57) or the placebo group (70 [95% CI, 67-72]; n = 87) (P = .18). Time to extubation was 17 minutes (95% CI, 14-20 minutes) in the lorazepam group, 12 minutes (95% CI, 11-13 minutes) for the no premedication group, and 13 minutes (95% CI, 12-14 minutes) for the placebo group (P < .001) and the rate of early cognitive recovery was 51% (95% CI, 45%-56%), 71% (95% CI, 66%-76%), and 64% (95% CI, 59%-69%), respectively (P < .001). Among patients undergoing elective surgery under general anesthesia, sedative premedication with lorazepam compared with placebo or no premedication did not improve the self-reported patient experience the day after surgery, but was associated with modestly prolonged time to extubation and a lower rate of early cognitive recovery. The findings suggest a lack of benefit with routine use of lorazepam as sedative premedication in patients undergoing general anesthesia. clinicaltrials.gov Identifier: NCT01901003.
Product Description
Supelco
Lorazepam solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Lorazepam for system suitability, European Pharmacopoeia (EP) Reference Standard