Merck
  • A study on exposure to cyanide among a community living near a gold mine in Malaysia.
A study on exposure to cyanide among a community living near a gold mine in Malaysia.
Journal of environmental health (2015-01-27)
Noor Artika Hassan, Mazrura Sahani, Rozita Hod, Noral' Asyikin Yahya
ABSTRACT
Cyanidation in gold extraction has detrimental impacts on the ecosystem and the population in surrounding areas. The research described in this article aimed to assess the health effects of environmental exposure to cyanide in a community near a gold mining area in Malaysia. A total of 255 members of the exposed community and 117 members of the control community answered questionnaires on their exposure to a gold mine and health outcomes. Urine samples were collected for biomarker assessment (i.e., urinary thiocyanate). The authors found that the mean level of urinary thiocyanate was significantly higher among the exposed community (0.30 ± 0.26 mg/dL) than the control community (0.24 ± 0.23 mg/dL) (p = .0021). The exposed community had a significantly higher prevalence of headaches (prevalence odds ratio [pOR] = 2.6, p = .001), dizziness (pOR = 3.7, p < .001), skin irritation (pOR = 2.1, p = .040), and eye irritation (pOR = 5.1, p = .004). No association occurred between dietary exposure and urinary biomarkers. Smokers living near a gold mine had a higher urinary thiocyanate level (0.56 mg/dL) than smokers in the control community (0.49 mg/dL).
Sigma-Aldrich
Gold, powder, <45 μm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, beads, 1-6 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, powder, <10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.997% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Gold, tube, 200mm, outside diameter 2.0mm, inside diameter 1.8mm, wall thickness 0.1mm, as drawn, 99.95%
Gold, tube, 100mm, outside diameter 1.0mm, inside diameter 0.5mm, wall thickness 0.25mm, as drawn, 99.95%
Gold, tube, 100mm, outside diameter 3.0mm, inside diameter 2.8mm, wall thickness 0.10mm, as drawn, 99.95%
Gold, rod, 25mm, diameter 4.0mm, as drawn, 99.95%
Gold, insulated wire, 0.5m, conductor diameter 0.125mm, insulation thickness 0.016mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Gold, insulated wire, 5m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyester insulation, 99.99%
Gold, microfoil, disks, 10mm, thinness 0.1μm, specific density 204.1μg/cm2, permanent mylar 3.5μm support, 99.99+%
Gold, tube, 50mm, outside diameter 1.0mm, inside diameter 0.5mm, wall thickness 0.25mm, as drawn, 99.95%
Gold, tube, 100mm, outside diameter 1.60mm, inside diameter 0.6mm, wall thickness 0.5mm, as drawn, 99.95%
Gold, microleaf disks, 10mm, thinness 0.5μm, specific density 966μg/cm2, removable support, 99.99+%
Gold, insulated wire, 0.5m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyester insulation, 99.99%
Gold, insulated wire, 1m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyester insulation, 99.99%
Gold, rod, 6mm, diameter 6.0mm, as drawn, 99.95%
Gold, tube, 100mm, outside diameter 10.0mm, inside diameter 9.7mm, wall thickness 0.15mm, as drawn, 99.95%
Gold, rod, 10mm, diameter 6.0mm, as drawn, 99.95%
Gold, microfoil, disks, 10mm, thinness 0.05μm, specific density 101.3μg/cm2, permanent mylar 3.5μm support, 99.99+%
Gold, rod, 100mm, diameter 2.0mm, as drawn, 99.95%