Merck
  • Dynamic high performance liquid chromatography on chiral stationary phases. Low temperature separation of the interconverting enantiomers of diazepam, flunitrazepam, prazepam and tetrazepam.
Dynamic high performance liquid chromatography on chiral stationary phases. Low temperature separation of the interconverting enantiomers of diazepam, flunitrazepam, prazepam and tetrazepam.
Journal of chromatography. A (2014-08-21)
Rocchina Sabia, Alessia Ciogli, Marco Pierini, Francesco Gasparrini, Claudio Villani
ABSTRACT
Diazepam and the structurally related 1,4-benzodiazepin-2-ones tetrazepam, prazepam and flunitrazepam are chiral molecules because they adopt a ground state conformation featuring a non-planar seven membered ring devoid of any reflection-symmetry element. The two conformational enantiomers of this class of benzodiazepines interconvert rapidly at room temperature by a simple ring flipping process. Low temperature HPLC on the Whelk-O1 chiral stationary phase allowed us to separate the conformational enantiomers of diazepam and of the related 1,4-benzodiazepin-2-ones, under conditions where the interconversion rate is sufficiently low, compared to the chromatographic separation rate. Diazepam, tetrazepam and prazepam showed temperature dependent dynamic HPLC profiles with interconversion plateaus indicative of on-column enantiomer interconversion (enantiomerization) in the temperature range between -10 °C and -35 °C, whereas for flunitrazepam on-column interconversion was observed at temperatures between -40 °C and -66 °C. Simulation of exchange-deformed HPLC profiles using a computer program based on the stochastic model yielded the apparent rate constants for the on-column enantiomerization and the corresponding free energy activation barriers. At -20 °C the enantiomerization barriers, ΔG(≠), for diazepam, prazepam and tetrazepam were determined to be in the range 17.6-18.7 kcal/mol. At -55 °C ΔG(≠) for flunitrazepam was determined to be in the 15.6-15.7 kcal/mol range. The experimental dynamic chromatograms and the corresponding interconversion barriers reported in this paper call for a reinterpretation of previously published results on the HPLC behavior of diazepam on chiral stationary phases.
Product Description
Diazepam for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, analytical standard
Supelco
Hexane, analytical standard
Supelco
Prazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, anhydrous, 95%
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Supelco
Diazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%