Merck
  • Surface expression of CD39 identifies an enriched Treg-cell subset in the rheumatic joint, which does not suppress IL-17A secretion.
Surface expression of CD39 identifies an enriched Treg-cell subset in the rheumatic joint, which does not suppress IL-17A secretion.
European journal of immunology (2014-07-06)
Jessica Herrath, Karine Chemin, Inka Albrecht, Anca I Catrina, Vivianne Malmström
ABSTRACT
Treg cells are important for the maintenance of self-tolerance and are implicated in autoimmunity. Despite enrichment of Treg cells in joints of rheumatoid arthritis (RA) patients, local inflammation persists. As expression of the ATP-hydrolyzing enzymes CD39 and CD73 and the resulting anti-inflammatory adenosine production have been implicated as an important mechanism of suppression, we characterized FOXP3(+) Treg cells in blood and synovial fluid samples of RA patients in the context of CD39 and CD73 expression. Synovial FOXP3(+) Treg cells displayed high expression levels of rate-limiting CD39, whereas CD73 was diminished. FOXP3(+) CD39(+) Treg cells were also abundant in synovial tissue. Furthermore, FOXP3(+) CD39(+) Treg cells did not secrete the proinflammatory cytokines IFN-γ and TNF after in vitro stimulation in contrast to FOXP3(+) CD39(-) T cells. FOXP3(+) CD39(+) Treg cells could be isolated by CD39 and CD25 coexpression, displayed a demethylated Treg-specific demethylated region and coculture assays confirmed that CD25(+) CD39(+) T cells have suppressive capacity, while their CD39(-) counterparts do not. Overall, our data show that FOXP3(+) CD39(+) Treg cells are enriched at the site of inflammation, do not produce proinflammatory cytokines, and are good suppressors of many effector T-cell functions including production of IFN-γ, TNF, and IL-17F but do not limit IL-17A secretion.
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥60 units/mg protein, lyophilized powder (partially purified)
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥3.0 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
Diethyl azodicarboxylate solution, purum, ~40% in toluene (H-NMR)
Sigma-Aldrich
Apyrase from potatoes, High Activity, ATPase ≥600 units/mg protein, lyophilized powder