Merck
  • Linking cellulose fiber sediment methyl mercury levels to organic matter decay and major element composition.
Linking cellulose fiber sediment methyl mercury levels to organic matter decay and major element composition.
Ambio (2014-01-15)
Olof Regnell, Mark Elert, Lars Olof Höglund, Anna Helena Falk, Anders Svensson
ABSTRACT
Methylation of mercury (Hg) to highly toxic methyl Hg (MeHg), a process known to occur when organic matter (OM) decomposition leads to anoxia, is considered a worldwide threat to aquatic ecosystems and human health. We measured temporal and spatial variations in sediment MeHg, total Hg (THg), and major elements in a freshwater lagoon in Sweden polluted with Hg-laden cellulose fibers. Fiber decomposition, confined to a narrow surface layer, resulted in loss of carbon (C), uptake of nitrogen (N), phosphorus (P), and sulfur (S), and increased MeHg levels. Notably, fiber decomposition and subsequent erosion of fiber residues will cause buried contaminants to gradually come closer to the sediment-water interface. At an adjacent site where decomposed fiber accumulated, there was a gain in C and a loss of S when MeHg increased. As evidenced by correlation patterns and vertical chemical profiles, reduced S may have fueled C-fixation and Hg methylation at this site.
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
α-Cellulose, powder
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Supelco
Cellulose, powder, for column chromatography
Supelco
Cellulose, acid washed, from spruce, for column chromatography
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Sigma-Aldrich
Cellulose, fibers, (medium)
Sigma-Aldrich
Cellulose, microcrystalline, powder
Supelco
Cellulose, acid washed, powder, for column chromatography
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Cellulose, colloidal, microcrystalline
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm