- Dissociative photoionization of methoxycarbonylsulfenyl chloride, CH3OC(O)SCl, following sulfur 2p, chlorine 2p, and oxygen 1s excitations.
Dissociative photoionization of methoxycarbonylsulfenyl chloride, CH3OC(O)SCl, following sulfur 2p, chlorine 2p, and oxygen 1s excitations.
Total ion yield spectra and photoinduced fragmentations following S 2p, Cl 2p, and O 1s inner shell excitations of methoxycarbonylsulfenyl chloride, CH(3)OC(O)SCl, have been studied in the gaseous phase by using synchrotron radiation and multicoincidence techniques, which include photoelectron-photoion coincidence (PEPICO) and photoelectron-photoion-photoion coincidence (PEPIPICO) time-of-flight (TOF) mass spectrometry. According to the analysis of the partial ion yield spectra the S+ ion signal shows a steep enhancement near the S 2p resonance, which could represent an evidence of state-specific fragmentations promoted by electronic excitations from the S 2p to vacant orbitals with strong antibonding character mainly located at the sulfur atom. The dissociation dynamics for selected PEPIPICO islands have been discussed. Fragmentation channels that involve the extrusion of H+ and CH(x)+ (x = 0, 1, 2, 3) fragments have been predominantly observed for dissociation of doubly charged CH(3)OC(O)SCl(2+).