Merck
  • Comparison of marginal fit of Lava CAD/CAM crown-copings with two finish lines.
Comparison of marginal fit of Lava CAD/CAM crown-copings with two finish lines.
The international journal of esthetic dentistry (2014-08-16)
Dino Re, Francesca Cerutti, Gabriele Augusti, Antonio Cerutti, Davide Augusti
ABSTRACT
Marginal fit is valued as one of the most important criteria for the clinical quality and success of all-ceramic crowns. The aim of this in vitro study was to investigate the marginal fit of Lava Zirconia crown-copings on chamfer and shoulder preparations. Two acrylic model teeth were selected to simulate the clinical preparations: one molar was prepared with a chamfer finish line (C) and one premolar was prepared with a rounded shoulder finish line (RS). Each resin model was duplicated 10 times using silicon-based impression material and poured in type IV dental stone for the fabrication of working dies. A total of 20 copings were divided into two groups (n = 10 for each finish line). Fifty measuring locations were chosen randomly along the margin on the dies and the gap width - vertical marginal discrepancy - was measured under a light microscope with a magnification of x100. Measurements were made without cementation. The mean marginal gap widths and standard deviations were calculated and a one-way analysis of variance (ANOVA) was performed for different types of preparations in order to detect differences (α = 0.05). The mean marginal gap was 30 ± 3 μm for the C group and 28 ± 4 μm for the RS group. The one-way ANOVA showed no statistical significant difference between the two groups (P = 0.23). Within the limitations of this study, the marginal discrepancies were all within the clinical acceptable standard set at 120 μm. Chamfer and shoulder preparations did not show differences regarding the gap dimension. Bur design is an easily selected parameter before natural tooth preparation. Both tested finish lines are able to help clinicians in obtaining acceptable marginal fit values for the investigated zirconia copings.
Sigma-Aldrich
Zirconium, powder, −100 mesh
Sigma-Aldrich
Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Zirconium, sponge, ≥99% trace metals basis
Sigma-Aldrich
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Zirconium, foil, 15mm disks, thickness 0.025mm, annealed, 99.2%
Zirconium, foil, 50x50mm, thickness 0.25mm, annealed, 99.2%
Zirconium, foil, not light tested, 25x25mm, thickness 0.003mm, 99.2%
Zirconium, foil, light tested, 50x50mm, thickness 0.015mm, 99.2%
Zirconium, foil, light tested, 25x25mm, thickness 0.006mm, 99.2%
Zirconium, foil, 8mm disks, thickness 0.25mm, annealed, 99.2%
Zirconium, foil, 8mm disks, thickness 0.5mm, annealed, 99.2%
Zirconium, foil, 6mm disks, thickness 0.075mm, annealed, 99.2%
Zirconium, foil, light tested, 25x25mm, thickness 0.01mm, 99.2%
Zirconium, foil, 4mm disks, thickness 0.01mm, 99.2%
Zirconium, foil, 4mm disks, thickness 0.25mm, annealed, 99.2%
Zirconium, foil, 6mm disks, thickness 0.25mm, annealed, 99.2%
Zirconium, foil, light tested, 50x50mm, thickness 0.05mm, annealed, 99.2%
Zirconium, foil, 50mm disks, thickness 0.25mm, annealed, 99.2%
Zirconium, foil, light tested, 25x25mm, thickness 0.01mm, 99.2%
Zirconium, foil, 6mm disks, thickness 0.01mm, 99.2%
Zirconium, foil, 50mm disks, thickness 0.016mm, 99.2%
Zirconium, foil, light tested, 50x50mm, thickness 0.008mm, 99.2%
Zirconium, foil, not light tested, 25x25mm, thickness 0.005mm, 99.2%
Zirconium, foil, light tested, 50x50mm, thickness 0.01mm, 99.2%
Zirconium, foil, light tested, 200x300mm, thickness 0.02mm, 99.2%
Zirconium, foil, 15mm disks, thickness 0.25mm, annealed, 99.2%
Zirconium, foil, 0.5m coil, thickness 0.5mm, annealed, 99.2%
Zirconium, foil, 300x300mm, thickness 0.25mm, annealed, 99.2%
Zirconium, foil, 0.5m coil, thickness 0.01mm, 99.2%
Zirconium, foil, 25mm disks, thickness 0.009mm, 99.2%