Merck
Genetic toxicology of 1,2-dibromo-3-chloropropane (DBCP).
Mutation research (1989-07-01)
S Teramoto, Y Shirasu
ABSTRACT
1,2-Dibromo-3-chloropropane (DBCP) is a nematocide, which has been used extensively as a soil fumigant in agriculture. Since sterility was found among male workers involved in the manufacture of DBCP, great concern has been focused on the genetic hazards of DBCP. DBCP gave positive results in many tests such as microbial, in vitro cytogenetics, and Drosophila studies. In mammalian test systems, DBCP caused chromosomal aberrations in the bone marrow cells and dominant-lethal mutations in germ cells in rats. In mice, there were no signs of DBCP-induced heritable mutation in germ cells, although point mutations were detected in somatic cells. The occurrence of Y-chromosomal non-disjunction was indicated in DBCP-exposed male workers by an increased number of sperm containing 2 Y-chromosomes.
Sigma-Aldrich
1,2-Dibromo-3-chloropropane, 97%