Merck
Fragrance material review on dl-citronellol.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2008-07-22)
A Lapczynski, S P Bhatia, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of dl-citronellol when used as a fragrance ingredient is presented.
Product Description
Supelco
(±)-β-Citronellol, analytical standard
Sigma-Aldrich
Citronellol, ≥95%, FCC, FG
Sigma-Aldrich
Citronellol, mixture of isomers, natural, ≥95%, FG
Sigma-Aldrich
β-Citronellol, 95%