Merck
  • Properties of olopatadine hydrochloride, a new antiallergic/antihistaminic drug.
Properties of olopatadine hydrochloride, a new antiallergic/antihistaminic drug.
Arzneimittel-Forschung (2005-01-14)
Kenji Ohmori, Kazuhide Hasegawa, Tadafumi Tamura, Kiyomi Miyake, Masahiro Matsubara, Shigehiro Masaki, Akira Karasawa, Naoko Urayama, Kaori Horikoshi, Jiro Kajita, Masanori Hasegawa, Koji Taniguchi, Toshio Komada, Yuji Kawamoto
ABSTRACT
Olopatadine hydrochloride (CAS 140462-76-6, KW-4679, AL-4943A; hereinafter referred to as olopatadine) is a novel antiallergic drug that is a selective histamine H1 receptor antagonist possessing inhibitory effects on the release of inflammatory lipid mediators such as leukotriene and thromboxane from human polymorphonuclear leukocytes and eosinophils. Olopatadine also inhibits the tachykininergic contractions in guinea pig bronchi by prejunctional inhibition of peripheral sensory nerves. Oral administration of olopatadine at doses of 0.03 mg/kg or higher reduces the symptoms of experimental allergic cutaneous responses and rhinoconjunctivitis in sensitized animals. Preclinical and clinical evaluations have demonstrated that olopatadine is a safe drug. After oral administration to healthy volunteers, olopatadine was rapidly and extensively absorbed. Unlike most other antiallergic drugs which are eliminated via hepatic metabolism, olopatadine is mainly excreted into urine. Olopatadine did not affect cytochrome P450 activities in human liver microsomes and consequently drug-drug metabolic interactions are unlikely. In double-masked clinical trials, olopatadine was shown to be effective at alleviating symptoms of allergic diseases. The drug (Allelock) was approved in Japan for the treatment of allergic rhinitis, chronic urticaria, eczema dermatitis, prurigo, cutaneous pruritus, psoriasis vulgaris and erythema exsudativum multiforme in December, 2000. An ophthalmic solution of olopatadine is also useful for the treatment of allergic conjunctivitis: this formulation (Patanol) was approved in the USA and the European Union for the treatment of seasonal and perennial allergic conjunctivitis in 1996 and 2002, respectively.
Sigma-Aldrich
Olopatadine hydrochloride, ≥98% (HPLC)
USP
Olopatadine hydrochloride, United States Pharmacopeia (USP) Reference Standard